Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_345258.7 | XP_345259.5 | 2 - 3 | 201 | 72 | |
GenBank mRNA | FQ221348.1 | - | 1 - 2 | 105 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Probe spans exons |