Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232996.2 | XP_006233058.1 | 3 - 4 | 215 | 101 | |
XM_345258.7 | XP_345259.5 | 5 - 6 | 485 | 101 | ||
GenBank mRNA | FQ221348.1 | - | 4 - 5 | 389 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |