Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106455.1 | NP_001099925.1 | 4 - 5 | 1249 | 64 | |
XM_006233041.2 | XP_006233103.1 | 6 - 7 | 2052 | 64 | ||
XM_006233042.2 | XP_006233104.1 | 6 - 7 | 2049 | 64 | ||
XM_006233043.2 | XP_006233105.1 | 5 - 6 | 1992 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |