Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024244.1 | NP_001019415.1 | 1 - 2 | 120 | 69 | |
GenBank mRNA | BC095842.1 | - | 1 - 2 | 120 | 7389454 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |