Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hormad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108949.1 | NP_001102419.1 | 11 - 12 | 941 | 124 | |
XM_006232951.2 | XP_006233013.1 | 10 - 11 | 896 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |