Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109655.2 | NP_001103125.1 | 9 - 10 | 863 | 92 | |
XM_006232988.2 | XP_006233050.1 | 10 - 11 | 1408 | 92 | ||
XM_006232989.2 | XP_006233051.1 | 10 - 11 | 1386 | 92 | ||
XM_006232990.2 | XP_006233052.1 | 10 - 11 | 1368 | 92 | ||
GenBank mRNA | EF187253.1 | - | 9 - 10 | 863 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |