Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133420.1 | NP_596911.1 | 1 - 2 | 239 | 88 | |
GenBank mRNA | L10077.1 | - | 1 - 2 | 239 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |