Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025142.2 | NP_001020313.2 | 2 - 3 | 582 | 66 | |
GenBank mRNA | BC085942.1 | - | 2 - 3 | 545 | 7126503 | 66 |
FQ224662.1 | - | 2 - 3 | 575 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |