Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gulo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022220.2 | NP_071556.2 | 3 - 4 | 261 | 68 | |
XM_006252146.2 | XP_006252208.1 | 3 - 4 | 402 | 68 | ||
GenBank mRNA | BC089803.1 | - | 3 - 4 | 261 | 7324344 | 68 |
FQ219140.1 | - | 3 - 4 | 286 | 68 | ||
FQ219632.1 | - | 3 - 4 | 369 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |