Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198764.1 | NP_942059.1 | 9 - 10 | 2160 | 79 | |
XM_008771225.1 | XP_008769447.1 | 8 - 9 | 2277 | 79 | ||
GenBank mRNA | AF515735.1 | - | 9 - 10 | 2160 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |