Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037181.1 | NP_001032258.1 | 5 - 6 | 731 | 65 | |
XM_006253087.2 | XP_006253149.1 | 4 - 5 | 560 | 65 | ||
XM_006253089.2 | XP_006253151.1 | 5 - 6 | 795 | 65 | ||
XM_006253090.2 | XP_006253152.1 | 3 - 4 | 419 | 65 | ||
XM_008771223.1 | XP_008769445.1 | 5 - 6 | 990 | 65 | ||
GenBank mRNA | BC107470.1 | - | 5 - 6 | 731 | 7457548 | 65 |
FQ228732.1 | - | 5 - 6 | 795 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |