Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsfl1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031981.2 | NP_114187.1 | 6 - 7 | 753 | 109 | |
XM_006235254.2 | XP_006235316.1 | 7 - 8 | 726 | 109 | ||
GenBank mRNA | AB002086.1 | - | 6 - 7 | 753 | 109 | |
BC072464.1 | - | 6 - 7 | 650 | 7096463 | 109 | |
FQ228953.1 | - | 7 - 8 | 691 | 109 | ||
Y10769.1 | - | 6 - 7 | 650 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |