Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsfl1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031981.2 | NP_114187.1 | 2 - 3 | 311 | 86 | |
XM_006235254.2 | XP_006235316.1 | 2 - 3 | 278 | 86 | ||
GenBank mRNA | AB002086.1 | - | 2 - 3 | 311 | 86 | |
BC072464.1 | - | 2 - 3 | 208 | 7096463 | 86 | |
FQ228953.1 | - | 2 - 3 | 243 | 86 | ||
Y10769.1 | - | 2 - 3 | 208 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |