Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109207.2 | NP_001102677.2 | 2 - 3 | 253 | 107 | |
GenBank mRNA | BC166989.1 | - | 2 - 3 | 253 | 9098732 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |