Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012580.2 | NP_036712.1 | 3 - 4 | 751 | 94 | |
GenBank mRNA | BC091164.1 | - | 3 - 4 | 751 | 7384448 | 94 |
FQ228696.1 | - | 2 - 3 | 625 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |