Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012040.1 | NP_001012040.1 | 2 - 3 | 804 | 79 | |
XM_006251640.1 | XP_006251702.1 | 2 - 3 | 842 | 79 | ||
GenBank mRNA | BC082100.1 | - | 2 - 3 | 804 | 7134467 | 79 |
FQ219794.1 | - | 2 - 3 | 842 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |