Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rarb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031529.1 | NP_113717.1 | 1 - 2 | 693 | 81 | |
GenBank mRNA | AJ002942.1 | - | 1 - 2 | 351 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Nuclear Receptor Recommended Assay Rattus norvegicus Inventoried Probe spans exons |