Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108440.1 | NP_001101910.1 | 14 - 15 | 1493 | 80 | |
XM_006255267.2 | XP_006255329.1 | 15 - 16 | 2028 | 80 | ||
XM_006255268.2 | XP_006255330.1 | 15 - 16 | 2025 | 80 | ||
XM_008772409.1 | XP_008770631.1 | 12 - 13 | 1634 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |