Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012057.1 | NP_001012057.1 | 3 - 4 | 462 | 62 | |
XM_008772405.1 | XP_008770627.1 | 4 - 5 | 583 | 62 | ||
XM_008772406.1 | XP_008770628.1 | 4 - 5 | 772 | 62 | ||
XM_008772407.1 | XP_008770629.1 | 4 - 5 | 509 | 62 | ||
GenBank mRNA | AY325245.1 | - | 532 | 62 | ||
FQ223845.1 | - | 658 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |