Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053847.1 | NP_446299.1 | 4 - 5 | 1088 | 68 | |
XM_006254048.2 | XP_006254110.1 | 4 - 5 | 1108 | 68 | ||
GenBank mRNA | M94454.1 | - | 4 - 5 | 1088 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |