Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053925.1 | NP_446377.1 | 357 | 106 | ||
GenBank mRNA | AB022014.1 | - | 357 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |