Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270712.1 | NP_001257641.1 | 1 - 2 | 98 | 82 | |
NM_130738.2 | NP_570094.1 | 1 - 2 | 98 | 82 | ||
GenBank mRNA | AF101041.1 | - | 1 - 2 | 58 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |