Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133285.1 | NP_579819.1 | 1 | 600 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |