Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177932.2 | NP_808793.1 | 7 - 8 | 766 | 147 | |
XM_006239127.2 | XP_006239189.1 | 8 - 9 | 1067 | 147 | ||
XM_008764144.1 | XP_008762366.1 | 8 - 9 | 1069 | 147 | ||
GenBank mRNA | AY225592.1 | - | 7 - 8 | 766 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |