Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198130.1 | NP_937763.1 | 4 - 5 | 819 | 121 | |
XM_006244087.2 | XP_006244149.1 | 5 - 6 | 1166 | 121 | ||
GenBank mRNA | AY191270.1 | - | 4 - 5 | 819 | 121 | |
BC097266.1 | - | 4 - 5 | 759 | 7459961 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Rat Chemokine Panel 1700 Inventoried Probe spans exons |