Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017175.2 | NP_058871.2 | 19 - 20 | 2624 | 147 | |
XM_006255245.2 | XP_006255307.1 | 19 - 20 | 3664 | 147 | ||
GenBank mRNA | BC061836.1 | - | 19 - 20 | 2525 | 5599989 | 147 |
D26180.1 | - | 19 - 20 | 2575 | 147 | ||
EV774518.1 | - | 2 - 3 | 244 | 147 | ||
FQ222217.1 | - | 19 - 20 | 2624 | 147 | ||
L35634.1 | - | 19 - 20 | 2467 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |