Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042354.1 | NP_001035813.1 | 1 - 2 | 144 | 59 | |
NM_001042356.1 | NP_001035815.1 | 1 - 2 | 144 | 59 | ||
NM_021739.2 | NP_068507.2 | 1 - 2 | 144 | 59 | ||
XM_006251170.2 | XP_006251232.1 | 1 - 2 | 243 | 59 | ||
XM_006251171.2 | XP_006251233.1 | 1 - 2 | 243 | 59 | ||
XM_006251172.2 | XP_006251234.1 | 1 - 2 | 243 | 59 | ||
XM_006251174.2 | XP_006251236.1 | 1 - 2 | 243 | 59 | ||
XM_006251176.2 | XP_006251238.1 | 1 - 2 | 224 | 59 | ||
XM_006251177.2 | XP_006251239.1 | 1 - 2 | 223 | 59 | ||
GenBank mRNA | CB616255.1 | - | 1 - 2 | 144 | 59 | |
M16112.1 | - | 1 - 2 | 124 | 59 | ||
X83375.1 | - | 1 - 2 | 106 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Protein Phosphatase Rattus norvegicus Probe spans exons |