Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053441.1 | NP_445893.1 | 2 - 3 | 503 | 97 | |
XM_006237602.1 | XP_006237664.1 | 3 - 4 | 651 | 97 | ||
GenBank mRNA | AF306546.1 | - | 2 - 3 | 503 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |