Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013092.1 | NP_037224.1 | 2 - 3 | 240 | 114 | |
GenBank mRNA | D38495.1 | - | 2 - 3 | 240 | 114 | |
U67908.1 | - | 2 - 3 | 205 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protease Rattus norvegicus 1700 Inventoried Probe spans exons |