Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133580.1 | NP_598264.1 | 4 - 5 | 299 | 66 | |
GenBank mRNA | BC061984.1 | - | 5 - 6 | 607 | 5599180 | 66 |
U18771.1 | - | 4 - 5 | 299 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |