Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thrsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012703.2 | NP_036835.2 | 1 | 345 | 64 | |
GenBank mRNA | BC089061.1 | - | 1 | 345 | 7308757 | 64 |
FQ214269.1 | - | 1 | 356 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |