Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030585.1 | NP_085074.1 | 7 - 8 | 1108 | 138 | |
GenBank mRNA | BC089795.1 | - | 7 - 8 | 1094 | 7375879 | 138 |
U34932.1 | - | 7 - 8 | 1108 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |