Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053955.1 | NP_446407.1 | 5 - 6 | 702 | 91 | |
GenBank mRNA | BC088121.1 | - | 5 - 6 | 733 | 7323247 | 91 |
Y17328.1 | - | 5 - 6 | 702 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |