Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107761.1 | NP_001101231.1 | 5 - 6 | 1184 | 74 | |
XM_006234766.2 | XP_006234828.1 | 5 - 6 | 945 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |