Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044301.1 | NP_001037766.1 | 2 - 3 | 829 | 68 | |
XM_006234695.2 | XP_006234757.1 | 3 - 4 | 1065 | 68 | ||
XM_008762107.1 | XP_008760329.1 | 2 - 3 | 842 | 68 | ||
XM_008762108.1 | XP_008760330.1 | 2 - 3 | 878 | 68 | ||
GenBank mRNA | BC083925.1 | - | 2 - 3 | 829 | 7135360 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |