Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008771.3 | NP_001008771.2 | 6 - 7 | 716 | 99 | |
GenBank mRNA | BC097367.1 | - | 6 - 7 | 667 | 7443204 | 99 |
FQ213175.1 | - | 6 - 7 | 811 | 99 | ||
FQ226328.1 | - | 6 - 7 | 711 | 99 |
Gene Symbol: LOC100911453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749373.3 | XP_003749421.1 | 6 - 7 | 822 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |