Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077645.1 | NP_001071113.1 | 2 - 3 | 167 | 124 | |
XM_006233451.2 | XP_006233513.1 | 2 - 3 | 884 | 124 | ||
XM_006233452.2 | XP_006233514.1 | 2 - 3 | 2389 | 124 | ||
XM_006233453.2 | XP_006233515.1 | 2 - 3 | 631 | 124 | ||
GenBank mRNA | D10770.1 | - | 2 - 3 | 167 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |