Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrtm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109430.1 | NP_001102900.1 | 3 - 4 | 511 | 70 | |
XM_006237208.2 | XP_006237270.1 | 4 - 5 | 1164 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |