Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017008.4 | NP_058704.1 | 1 - 2 | 54 | 60 | |
GenBank mRNA | BC059110.1 | - | 1 - 2 | 55 | 6919611 | 60 |
X02231.1 | - | 1 - 2 | 50 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |