Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107898.1 | NP_001101368.1 | 3 - 4 | 1910 | 114 | |
XM_006237743.2 | XP_006237805.1 | 4 - 5 | 2001 | 114 | ||
XM_008763495.1 | XP_008761717.1 | 4 - 5 | 2307 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |