Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013936.1 | NP_001013958.1 | 2 - 3 | 589 | 67 | |
GenBank mRNA | BC083723.1 | - | 2 - 3 | 589 | 7191003 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |