Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106701.2 | NP_001100171.1 | 1 - 2 | 60 | 65 | |
GenBank mRNA | BC158867.1 | - | 1 - 2 | 60 | 5623763 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |