Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108667.1 | NP_001102137.1 | 2 - 3 | 441 | 109 | |
XM_006238166.2 | XP_006238228.1 | 4 - 5 | 548 | 109 | ||
XM_006238168.2 | XP_006238230.1 | 3 - 4 | 475 | 109 | ||
XM_006238169.1 | XP_006238231.1 | 2 - 3 | 358 | 109 | ||
XM_008763712.1 | XP_008761934.1 | 3 - 4 | 568 | 109 | ||
XM_008763713.1 | XP_008761935.1 | 4 - 5 | 542 | 109 | ||
XM_008763714.1 | XP_008761936.1 | 3 - 4 | 476 | 109 | ||
XM_008763715.1 | XP_008761937.1 | 3 - 4 | 475 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |