Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108667.1 | NP_001102137.1 | 3 - 4 | 498 | 101 | |
XM_006238166.2 | XP_006238228.1 | 5 - 6 | 605 | 101 | ||
XM_006238168.2 | XP_006238230.1 | 4 - 5 | 532 | 101 | ||
XM_006238169.1 | XP_006238231.1 | 3 - 4 | 415 | 101 | ||
XM_008763712.1 | XP_008761934.1 | 4 - 5 | 625 | 101 | ||
XM_008763713.1 | XP_008761935.1 | 5 - 6 | 599 | 101 | ||
XM_008763714.1 | XP_008761936.1 | 4 - 5 | 533 | 101 | ||
XM_008763715.1 | XP_008761937.1 | 4 - 5 | 532 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |