Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108667.1 | NP_001102137.1 | 6 - 7 | 1107 | 79 | |
XM_006238166.2 | XP_006238228.1 | 8 - 9 | 1214 | 79 | ||
XM_006238168.2 | XP_006238230.1 | 7 - 8 | 1141 | 79 | ||
XM_006238169.1 | XP_006238231.1 | 6 - 7 | 1024 | 79 | ||
XM_008763712.1 | XP_008761934.1 | 7 - 8 | 1234 | 79 | ||
XM_008763713.1 | XP_008761935.1 | 8 - 9 | 1208 | 79 | ||
XM_008763714.1 | XP_008761936.1 | 7 - 8 | 1142 | 79 | ||
XM_008763715.1 | XP_008761937.1 | 7 - 8 | 1141 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |