Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134619.1 | NP_001128091.1 | 2 - 3 | 2532 | 123 | |
XM_006235428.2 | XP_006235490.1 | 3 - 4 | 2609 | 123 | ||
XM_008762375.1 | XP_008760597.1 | 3 - 4 | 3372 | 123 | ||
GenBank mRNA | FQ216244.1 | - | 2 - 3 | 245 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |