Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199082.2 | NP_954513.2 | 2 - 3 | 211 | 61 | |
XM_006247879.2 | XP_006247941.1 | 2 - 3 | 307 | 61 | ||
XM_006247880.2 | XP_006247942.1 | 3 - 4 | 1131 | 61 | ||
XM_006247881.2 | XP_006247943.1 | 2 - 3 | 848 | 61 | ||
XM_006247882.2 | XP_006247944.1 | 2 - 3 | 304 | 61 | ||
XM_006247884.2 | XP_006247946.1 | 3 - 4 | 1129 | 61 | ||
GenBank mRNA | DY309177.1 | - | 1 - 2 | 299 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |