Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myocd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182667.2 | NP_872608.1 | 11 - 12 | 2433 | 90 | |
XM_006246582.2 | XP_006246644.1 | 12 - 13 | 5018 | 90 | ||
XM_006246583.2 | XP_006246645.1 | 13 - 14 | 5062 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Rattus norvegicus Probe spans exons |