Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gntl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015035.1 | NP_001015035.1 | 11 - 12 | 1100 | 74 | |
XM_008768505.1 | XP_008766727.1 | 11 - 12 | 1150 | 74 | ||
XM_008768506.1 | XP_008766728.1 | 11 - 12 | 1150 | 74 | ||
XM_008768507.1 | XP_008766729.1 | 11 - 12 | 1150 | 74 | ||
XM_008768508.1 | XP_008766730.1 | 11 - 12 | 1150 | 74 | ||
GenBank mRNA | AY634365.1 | - | 11 - 12 | 1100 | 74 | |
BC111711.1 | - | 12 - 13 | 1149 | 7383845 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |