Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106985.1 | NP_001100455.1 | 1 - 2 | 228 | 58 | |
XM_006245947.2 | XP_006246009.2 | 1 - 2 | 228 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |